Product Details

SNP ID
rs199559680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152168501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTTGGCCAGGTTGATGGGATA[C/G]GTGGTCCCCACGATGTTGAAGGTAG
Phenotype
MIM: 305660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GABRA3 PubMed Links

Gene Details

Gene
GABRA3
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000808.3 1234 Silent Mutation ACC,ACG T402T NP_000799.1
XM_006724811.2 1234 Missense Mutation CTA,GTA L332V XP_006724874.1

View Full Product Details