Product Details

SNP ID
rs200059622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49176078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCGGCGCGGGCCTCCTTCAGA[C/T]ACCAGAGGGTCGCGGAAGCTGACGC
Phenotype
MIM: 300112 MIM: 300111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGIX PubMed Links

Gene Details

Gene
MAGIX
Gene Name
MAGI family member, X-linked
There are no transcripts associated with this gene.

Gene
PLP2
Gene Name
proteolipid protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002668.2 1546 Intron NP_002659.1
Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307979.1 1546 Silent Mutation GTA,GTG V413V NP_001294908.1
NM_006150.4 1546 Silent Mutation GTA,GTG V481V NP_006141.2

View Full Product Details