Product Details

SNP ID
rs200167143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103586976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCAGAGATAGAGGGAAAGCCA[C/G]AGAGTGAAGGAGAGCCAGGGAGTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL4 PubMed Links

Gene Details

Gene
TCEAL4
Gene Name
transcription elongation factor A like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006935.2 595 Missense Mutation CAG,GAG Q101E NP_001006936.1
NM_001006937.2 595 Missense Mutation CAG,GAG Q101E NP_001006938.1
NM_001300901.1 595 Missense Mutation CAG,GAG Q244E NP_001287830.1
NM_001305840.1 595 Missense Mutation CAG,GAG Q101E NP_001292769.1
NM_001305841.1 595 Missense Mutation CAG,GAG Q101E NP_001292770.1
NM_001305842.1 595 Missense Mutation CAG,GAG Q101E NP_001292771.1
NM_024863.5 595 Missense Mutation CAG,GAG Q101E NP_079139.4
XM_017029856.1 595 Missense Mutation CAG,GAG Q163E XP_016885345.1
XM_017029857.1 595 Missense Mutation CAG,GAG Q101E XP_016885346.1

View Full Product Details