Product Details

SNP ID
rs201263889
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152168297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGTGGCCCAATAGACCAGATT[G/T]AATATGGCAAAGAGCACAGGAAAGA
Phenotype
MIM: 305660
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GABRA3 PubMed Links

Gene Details

Gene
GABRA3
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000808.3 1438 Missense Mutation TTA,TTC L470F NP_000799.1
XM_006724811.2 1438 UTR 3 XP_006724874.1

View Full Product Details