Product Details

SNP ID
rs201030059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:17800833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACTATCCATCATGGCTGGGGCC[G/T]CATGGGACGTGGCAGGGGCAAAAGA
Phenotype
MIM: 300217
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAI2 PubMed Links

Gene Details

Gene
RAI2
Gene Name
retinoic acid induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172732.1 1823 Missense Mutation GAG,GCG E343A NP_001166203.1
NM_001172739.1 1823 Missense Mutation GAG,GCG E393A NP_001166210.1
NM_001172743.1 1823 Missense Mutation GAG,GCG E393A NP_001166214.1
NM_021785.4 1823 Missense Mutation GAG,GCG E393A NP_068557.3
XM_006724459.2 1823 Missense Mutation GAG,GCG E393A XP_006724522.1
XM_006724460.1 1823 Missense Mutation GAG,GCG E393A XP_006724523.1
XM_011545439.2 1823 Missense Mutation GAG,GCG E393A XP_011543741.1
XM_011545440.2 1823 Missense Mutation GAG,GCG E393A XP_011543742.1
XM_011545441.2 1823 Missense Mutation GAG,GCG E393A XP_011543743.1

View Full Product Details