Product Details

SNP ID
rs202046647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13040812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATTCTGTTCTTTATTTCGCTG[C/T]AAGCTTTTGCAATCTTTTCAATCCT
Phenotype
MIM: 300479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM9C PubMed Links

Gene Details

Gene
FAM9C
Gene Name
family with sequence similarity 9 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174901.5 602 Missense Mutation TAC,TGC Y92C NP_777561.1
XM_005274460.2 602 Intron XP_005274517.1
XM_017029321.1 602 Intron XP_016884810.1
Gene
LOC105373133
Gene Name
uncharacterized LOC105373133
There are no transcripts associated with this gene.

View Full Product Details