Product Details

SNP ID
rs201224415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129908690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGCGGAAATTGGCTGAGAGGT[C/T]TGAGGCTAGTCTGAAGGTGTCAGAG
Phenotype
MIM: 300508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP14A PubMed Links

Gene Details

Gene
UTP14A
Gene Name
UTP14A small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166221.1 284 Missense Mutation TCT,TTT S65F NP_001159693.1
NM_006649.3 284 Missense Mutation TCT,TTT S65F NP_006640.2
XM_017029237.1 284 UTR 5 XP_016884726.1
XM_017029238.1 284 UTR 5 XP_016884727.1

View Full Product Details