Product Details
- SNP ID
-
rs200599080
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:78657269 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCTGATGCACTGAGTGATGAG[A/T]TCAGATAGGTTGGTGGCTGGGCTTG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZCCHC5
PubMed Links
Gene Details
- Gene
- ZCCHC5
- Gene Name
- zinc finger CCHC-type containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152694.2 |
1448 |
Missense Mutation |
GAA,GAT |
E384D |
NP_689907.1 |
View Full Product Details