Product Details

SNP ID
rs201883808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154349534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTACAAACACTGATGATGTCTC[A/G]TGGAGGCTGTGGTTGCTGACGAGAC
Phenotype
MIM: 300017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLNA PubMed Links

Gene Details

Gene
FLNA
Gene Name
filamin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110556.1 7630 Silent Mutation CAC,CAT H2528H NP_001104026.1
NM_001456.3 7630 Silent Mutation CAC,CAT H2520H NP_001447.2
XM_011531127.1 7630 Silent Mutation CAC,CAT H2496H XP_011529429.1
XM_011531128.1 7630 Silent Mutation CAC,CAT H2488H XP_011529430.1
XM_011531129.1 7630 Silent Mutation CAC,CAT H2470H XP_011529431.1
XM_011531130.1 7630 Silent Mutation CAC,CAT H2462H XP_011529432.1
XM_011531131.1 7630 Silent Mutation CAC,CAT H2461H XP_011529433.1
Gene
LOC105373386
Gene Name
uncharacterized LOC105373386
There are no transcripts associated with this gene.

View Full Product Details