Product Details

SNP ID
rs6024830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56363151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCCTCCCCTCCAGAGCCTGGAC[A/G]TTCTGGAAAAGTGTCTTCCTTTAAG
Phenotype
MIM: 603072
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
There are no transcripts associated with this gene.

Gene
FAM210B
Gene Name
family with sequence similarity 210 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080821.2 Intron NP_543011.2

View Full Product Details