Product Details
- SNP ID
-
rs453202
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:127519813 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTGTCTCTACTTAGATGTTTGTT[G/T]CAAGGAACTTAAACAGCATGTCAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRRC1
PubMed Links
Gene Details
- Gene
- PRRC1
- Gene Name
- proline rich coiled-coil 1
View Full Product Details