Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014326.3 | 4097 | UTR 3 | NP_055141.2 | ||
XM_011521413.2 | 4097 | UTR 3 | XP_011519715.1 | ||
XM_011521414.1 | 4097 | UTR 3 | XP_011519716.1 | ||
XM_011521415.2 | 4097 | Intron | XP_011519717.1 | ||
XM_011521416.2 | 4097 | Intron | XP_011519718.1 | ||
XM_011521417.1 | 4097 | UTR 3 | XP_011519719.1 | ||
XM_011521421.2 | 4097 | UTR 3 | XP_011519723.1 | ||
XM_017022043.1 | 4097 | Intron | XP_016877532.1 | ||
XM_017022044.1 | 4097 | UTR 3 | XP_016877533.1 | ||
XM_017022045.1 | 4097 | UTR 3 | XP_016877534.1 | ||
XM_017022046.1 | 4097 | Intron | XP_016877535.1 | ||
XM_017022047.1 | 4097 | Intron | XP_016877536.1 | ||
XM_017022048.1 | 4097 | Intron | XP_016877537.1 | ||
XM_017022049.1 | 4097 | Intron | XP_016877538.1 | ||
XM_017022050.1 | 4097 | Intron | XP_016877539.1 | ||
XM_017022051.1 | 4097 | Intron | XP_016877540.1 | ||
XM_017022052.1 | 4097 | Intron | XP_016877541.1 |