Product Details

SNP ID
rs11859267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29962957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATTTATCCAGCAGTGCAGGGA[G/A]TTGAACTTTTGGAGTCGGGAACCTT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TMEM219 PubMed Links

Gene Details

Gene
TMEM219
Gene Name
transmembrane protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083613.1 Intron NP_001077082.1
NM_194280.3 Intron NP_919256.1
XM_005255123.1 Intron XP_005255180.1

View Full Product Details