Product Details

SNP ID
rs35282564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56361648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTCAATTACTTTTCAACATT[C/T]TAAAAACTTTCCGTTATGTAAAATA
Phenotype
MIM: 603243 MIM: 139311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMFR PubMed Links

Gene Details

Gene
AMFR
Gene Name
autocrine motility factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144.5 3023 UTR 3 NP_001135.3
NM_001323511.1 3023 UTR 3 NP_001310440.1
NM_001323512.1 3023 UTR 3 NP_001310441.1
XM_005255890.3 3023 UTR 3 XP_005255947.1
Gene
GNAO1
Gene Name
G protein subunit alpha o1
There are no transcripts associated with this gene.

View Full Product Details