Product Details

SNP ID
rs12605480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14072283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACACGTTTTTAAGTTACAGTA[C/G]CAAGATATTATCGTTGCCTGTGTTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF519 PubMed Links

Gene Details

Gene
ZNF519
Gene Name
zinc finger protein 519
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145287.3 15421 Intron NP_660330.2
XM_011525605.2 15421 Intron XP_011523907.1
XM_017025562.1 15421 UTR 3 XP_016881051.1
XM_017025563.1 15421 UTR 3 XP_016881052.1
XM_017025564.1 15421 UTR 3 XP_016881053.1
XM_017025565.1 15421 UTR 3 XP_016881054.1
XM_017025566.1 15421 Intron XP_016881055.1

View Full Product Details