Product Details

SNP ID
rs12645115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82486269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAGCTATTTTAAAAAAAAATC[A/C]CTGCAAATGGTGAGCCAAGACCGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105377310 PubMed Links

Gene Details

Gene
LOC105377310
Gene Name
syncytin-1-like
There are no transcripts associated with this gene.

Gene
RPS6P6
Gene Name
ribosomal protein S6 pseudogene
There are no transcripts associated with this gene.

Gene
TMEM150C
Gene Name
transmembrane protein 150C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080506.1 Intron NP_001073975.1

View Full Product Details