Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171815.1 | 828 | UTR 3 | NP_001165286.1 | ||
NM_001171816.1 | 828 | UTR 3 | NP_001165287.1 | ||
NM_178841.3 | 828 | UTR 3 | NP_849163.1 | ||
XM_011522845.2 | 828 | UTR 3 | XP_011521147.1 | ||
XM_011522846.1 | 828 | UTR 3 | XP_011521148.1 | ||
XM_011522847.1 | 828 | UTR 3 | XP_011521149.1 | ||
XM_017022910.1 | 828 | UTR 3 | XP_016878399.1 | ||
XM_017022911.1 | 828 | Intron | XP_016878400.1 | ||
XM_017022912.1 | 828 | Intron | XP_016878401.1 | ||
XM_017022913.1 | 828 | UTR 3 | XP_016878402.1 |