Product Details

SNP ID
rs36022414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37829756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGATTTCTCCAAAGGTGCTTA[C/T]CTTGACAGGAAAGAAAAGAACACTT
Phenotype
MIM: 172000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGM2 PubMed Links

Gene Details

Gene
PGM2
Gene Name
phosphoglucomutase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018290.3 Intron NP_060760.2
XM_011513711.2 Intron XP_011512013.1

View Full Product Details