Product Details
- SNP ID
-
rs10089937
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:70052164 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CACAGATGCTGCATGAGCAGCCATC[A/G]TCCTCCTTGTGTGAACGCCGGACAT
- Phenotype
-
MIM: 611781
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM14
PubMed Links
Gene Details
- Gene
- PRDM14
- Gene Name
- PR domain 14
View Full Product Details