Product Details
- SNP ID
-
rs998579
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:64150284 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCTATTACCTTTGGTGGCCAAATGC[T/A]GAAAGAAAAGCTGAGAGAGTAAGCT
- Phenotype
-
- Polymorphism
- T/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNHG25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147227884] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNHG25
- Gene Name
- small nucleolar RNA host gene 25
There are no transcripts associated with this gene.
- Gene
- SNORA50C
- Gene Name
- small nucleolar RNA, H/ACA box 50C
There are no transcripts associated with this gene.
- Gene
- SNORD104
- Gene Name
- small nucleolar RNA, C/D box 104
There are no transcripts associated with this gene.
- Gene
- TEX2
- Gene Name
- testis expressed 2
View Full Product Details