Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000282.3 | Intron | NP_000273.2 | |||
NM_001127692.2 | Intron | NP_001121164.1 | |||
NM_001178004.1 | Intron | NP_001171475.1 | |||
XM_005254059.2 | Intron | XP_005254116.1 | |||
XM_011521093.1 | Intron | XP_011519395.1 | |||
XM_017020605.1 | Intron | XP_016876094.1 | |||
XM_017020606.1 | Intron | XP_016876095.1 | |||
XM_017020607.1 | Intron | XP_016876096.1 | |||
XM_017020608.1 | Intron | XP_016876097.1 | |||
XM_017020609.1 | Intron | XP_016876098.1 | |||
XM_017020610.1 | Intron | XP_016876099.1 | |||
XM_017020611.1 | Intron | XP_016876100.1 | |||
XM_017020612.1 | Intron | XP_016876101.1 | |||
XM_017020613.1 | Intron | XP_016876102.1 | |||
XM_017020614.1 | Intron | XP_016876103.1 | |||
XM_017020615.1 | Intron | XP_016876104.1 | |||
XM_017020616.1 | Intron | XP_016876105.1 | |||
XM_017020617.1 | Intron | XP_016876106.1 | |||
XM_017020618.1 | Intron | XP_016876107.1 |