Product Details
- SNP ID
-
rs6974683
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142244795 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTCTGGGCTCAGGAAATGATGTAGC[C/T]GCGACCTCTACCAAGCCCTGCCTGT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MOXD2P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75387311] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MOXD2P
- Gene Name
- monooxygenase, DBH-like 2, pseudogene
There are no transcripts associated with this gene.
- Gene
- PRSS58
- Gene Name
- protease, serine 58
There are no transcripts associated with this gene.
View Full Product Details