Product Details

SNP ID
rs1129626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5205707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTTTTGCTCCCAAACTGCCCCA[C/T]AGTCCCAGGGGGAAAGGGTCCCTGA
Phenotype
MIM: 601576
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRS PubMed Links

Gene Details

Gene
PTPRS
Gene Name
protein tyrosine phosphatase, receptor type S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002850.3 7077 UTR 3 NP_002841.3
NM_130853.2 7077 UTR 3 NP_570923.2
NM_130854.2 7077 UTR 3 NP_570924.2
NM_130855.2 7077 UTR 3 NP_570925.2
XM_005259600.2 7077 UTR 3 XP_005259657.1
XM_005259606.2 7077 UTR 3 XP_005259663.1
XM_005259607.2 7077 UTR 3 XP_005259664.1
XM_005259609.1 7077 UTR 3 XP_005259666.1
XM_005259610.1 7077 UTR 3 XP_005259667.1
XM_011528157.2 7077 UTR 3 XP_011526459.1
XM_011528158.2 7077 UTR 3 XP_011526460.1
XM_017027065.1 7077 UTR 3 XP_016882554.1
XM_017027066.1 7077 UTR 3 XP_016882555.1
XM_017027067.1 7077 UTR 3 XP_016882556.1
XM_017027068.1 7077 UTR 3 XP_016882557.1
XM_017027069.1 7077 UTR 3 XP_016882558.1
XM_017027070.1 7077 UTR 3 XP_016882559.1
XM_017027071.1 7077 UTR 3 XP_016882560.1
XM_017027072.1 7077 UTR 3 XP_016882561.1
XM_017027073.1 7077 UTR 3 XP_016882562.1
XM_017027074.1 7077 UTR 3 XP_016882563.1
XM_017027075.1 7077 UTR 3 XP_016882564.1
XM_017027076.1 7077 UTR 3 XP_016882565.1

View Full Product Details