Product Details

SNP ID
rs1105561
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44165807 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGGAGGAGGCAGCAGAACCCCA[C/T]GTGAGTAAGACTTGTTGTTATTCTT
Phenotype
MIM: 604750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF226 PubMed Links

Gene Details

Gene
ZNF226
Gene Name
zinc finger protein 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032372.1 259 UTR 5 NP_001027544.1
NM_001032373.1 259 UTR 5 NP_001027545.1
NM_001032374.1 259 UTR 5 NP_001027546.1
NM_001146220.2 259 UTR 5 NP_001139692.1
NM_001319088.1 259 UTR 5 NP_001306017.1
NM_001319089.1 259 UTR 5 NP_001306018.1
NM_001319090.1 259 UTR 5 NP_001306019.1
NM_015919.3 259 UTR 5 NP_057003.2
NM_016444.2 259 UTR 5 NP_057528.2
XM_005259227.2 259 UTR 5 XP_005259284.1
XM_006723367.3 259 UTR 5 XP_006723430.1
XM_006723368.3 259 UTR 5 XP_006723431.1
XM_006723369.2 259 UTR 5 XP_006723432.1
XM_017027262.1 259 UTR 5 XP_016882751.1
XM_017027263.1 259 UTR 5 XP_016882752.1
XM_017027264.1 259 UTR 5 XP_016882753.1
XM_017027265.1 259 UTR 5 XP_016882754.1
Gene
ZNF234
Gene Name
zinc finger protein 234
There are no transcripts associated with this gene.

View Full Product Details