Product Details
- SNP ID
-
rs14693
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:31276306 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGGATGCTCACAACTAAATCCC[G/T]TTTACACAAAGTTCCAAACACTTAC
- Phenotype
-
MIM: 614119
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSHZ3
PubMed Links
Gene Details
- Gene
- TSHZ3
- Gene Name
- teashirt zinc finger homeobox 3
There are no transcripts associated with this gene.
View Full Product Details