Product Details

SNP ID
rs1801526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124746951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGGAAGAGAAGTCCCTATCCCA[A/C]ACCTGCCTGTCACGTTCCCTCCCCT
Phenotype
MIM: 614281 MIM: 602350 MIM: 606011
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ESAM PubMed Links

Gene Details

Gene
ESAM
Gene Name
endothelial cell adhesion molecule
There are no transcripts associated with this gene.

Gene
NRGN
Gene Name
neurogranin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126181.1 960 UTR 3 NP_001119653.1
NM_006176.2 960 UTR 3 NP_006167.1
Gene
VSIG2
Gene Name
V-set and immunoglobulin domain containing 2
There are no transcripts associated with this gene.

View Full Product Details