Product Details

SNP ID
rs1802719
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:151067611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTCCCTACTGTGTTGGTGGTG[C/T]TGATGAATCTGCCAGAGTTGAGTTC
Phenotype
MIM: 604684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42SE1 PubMed Links

Gene Details

Gene
CDC42SE1
Gene Name
CDC42 small effector 1
There are no transcripts associated with this gene.

Gene
GABPB2
Gene Name
GA binding protein transcription factor beta subunit 2
There are no transcripts associated with this gene.

Gene
MLLT11
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006818.3 1272 UTR 3 NP_006809.1

View Full Product Details