Product Details

SNP ID
rs1789221
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50881915 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGTCTTGTAGTTTAACGTTCCCA[A/T]TTTACTGATGAGAAAATGCAAATTG
Phenotype
MIM: 154270
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ME2 PubMed Links
Additional Information
For this assay, SNP(s) [rs78099802] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ME2
Gene Name
malic enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168335.1 Intron NP_001161807.1
NM_002396.4 Intron NP_002387.1

View Full Product Details