Product Details

SNP ID
rs6490473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30623546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTTGGTCCTTGTCTATGGTTTC[C/T]GTTACAGAGCTAAAAGTCTTGGAAT
Phenotype
MIM: 163905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGB1 PubMed Links

Gene Details

Gene
HMGB1
Gene Name
high mobility group box 1
There are no transcripts associated with this gene.

Gene
USPL1
Gene Name
ubiquitin specific peptidase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321532.1 Intron NP_001308461.1
NM_001321533.1 Intron NP_001308462.1
NM_001321534.1 Intron NP_001308463.1
NM_005800.4 Intron NP_005791.3
XM_005266213.3 Intron XP_005266270.1
XM_006719751.3 Intron XP_006719814.1
XM_017020314.1 Intron XP_016875803.1
XM_017020315.1 Intron XP_016875804.1
XM_017020316.1 Intron XP_016875805.1

View Full Product Details