Product Details

SNP ID
rs13314659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48407742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGAGTCAGGATTCGCGCCTTCT[C/T]GACAACTCCCCATGGGGGAACCCTT
Phenotype
MIM: 601053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNB1 PubMed Links

Gene Details

Gene
PLXNB1
Gene Name
plexin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130082.2 Intron NP_001123554.1
NM_002673.5 Intron NP_002664.2
XM_011533833.2 Intron XP_011532135.1
XM_011533834.1 Intron XP_011532136.1
XM_011533835.1 Intron XP_011532137.1
XM_011533836.1 Intron XP_011532138.1
XM_011533837.2 Intron XP_011532139.1
XM_017006630.1 Intron XP_016862119.1
XM_017006631.1 Intron XP_016862120.1

View Full Product Details