Product Details

SNP ID
rs35117343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101726282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTAGTAACATTTAAACAATAGT[A/G]TCATTACTTAGGCACAACAAATGGA
Phenotype
MIM: 615864
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP97 PubMed Links

Gene Details

Gene
CEP97
Gene Name
centrosomal protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303401.1 Intron NP_001290330.1
NM_024548.3 Intron NP_078824.2
XM_006713743.3 Intron XP_006713806.1
XM_011513127.1 Intron XP_011511429.1
XM_017007178.1 Intron XP_016862667.1

View Full Product Details