Product Details
- SNP ID
-
rs1055745
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:73223778 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTCCGGTGTACGGTGGCAGGCTAAC[A/G]GAGGTGACCAGGTCATCATTGGTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KHDC1L
PubMed Links
Gene Details
- Gene
- KHDC1L
- Gene Name
- KH domain containing 1 like
View Full Product Details