Product Details

SNP ID
rs11593903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26956816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGTACCATTTCCCTTATGCAAA[C/T]AGAATTATTTGATGGAAATGCTGTC
Phenotype
MIM: 610855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD26 PubMed Links

Gene Details

Gene
ANKRD26
Gene Name
ankyrin repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256053.1 Intron NP_001242982.1
NM_014915.2 Intron NP_055730.2
XM_006717423.2 Intron XP_006717486.1
XM_006717425.3 Intron XP_006717488.1
XM_011519416.2 Intron XP_011517718.1
XM_017015928.1 Intron XP_016871417.1
XM_017015929.1 Intron XP_016871418.1
XM_017015930.1 Intron XP_016871419.1
XM_017015931.1 Intron XP_016871420.1
XM_017015932.1 Intron XP_016871421.1
XM_017015933.1 Intron XP_016871422.1
XM_017015934.1 Intron XP_016871423.1

View Full Product Details