Product Details

SNP ID
rs11718768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13952733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGTGGGTACAGAGTGGGACTA[C/G]ACGGGAAAGAAGGAGGGGATGAAGG
Phenotype
MIM: 611167
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPRXL PubMed Links

Gene Details

Gene
TPRXL
Gene Name
tetrapeptide repeat homeobox like
There are no transcripts associated with this gene.

View Full Product Details