Product Details

SNP ID
rs12294299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43686010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGTATGAGAAAATACTGGAAAG[A/T]TACTAAGGTGCTTAACAAAGCAAAG
Phenotype
MIM: 609574
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B12 PubMed Links

Gene Details

Gene
HSD17B12
Gene Name
hydroxysteroid 17-beta dehydrogenase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016142.2 Intron NP_057226.1
XM_011520156.1 Intron XP_011518458.1
XM_017017881.1 Intron XP_016873370.1

View Full Product Details