Product Details

SNP ID
hCV11612409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101353959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACTGGAAATTTGGAGCCTACT[C/G]AGCTTGTGTATTCATCATCCAGGAC
Phenotype
MIM: 300300 MIM: 300356
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000061.2 1854 Nonsense Mutation TCA,TGA S554* NP_000052.1
NM_001287344.1 1854 Nonsense Mutation TCA,TGA S588* NP_001274273.1
NM_001287345.1 1854 Nonsense Mutation TCA,TGA S378* NP_001274274.1
Gene
TIMM8A
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
There are no transcripts associated with this gene.

View Full Product Details