Product Details
- SNP ID
-
rs12745115
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:161316554 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATTAAAATGGAGTCTCTTATGTCT[A/G]CTTTCTATACGGACACCTTAACAAT
- Phenotype
-
MIM: 159440
MIM: 602413
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MPZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114534259] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MPZ
- Gene Name
- myelin protein zero
There are no transcripts associated with this gene.
- Gene
- SDHC
- Gene Name
- succinate dehydrogenase complex subunit C
View Full Product Details