Product Details
- SNP ID
-
rs844
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
53
- Location
-
Chr.1:161677743 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- AGCCACAGACAATATGGTCCCAAAT[A/G]ACCGACTGCACCTTCTGTGCTTCAG
- Phenotype
-
MIM: 604590
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FCGR2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60519172] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCGR2B
- Gene Name
- Fc fragment of IgG receptor IIb
- Gene
- RPL31P11
- Gene Name
- ribosomal protein L31 pseudogene 11
There are no transcripts associated with this gene.
View Full Product Details