Product Details
- SNP ID
-
rs4652540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:180952872 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGGAAATGGGAGGGGGCTTTGG[C/T]TCCTCCAGCCTCCTGTTTGACCTTC
- Phenotype
-
MIM: 603944
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1614
PubMed Links
Gene Details
- Gene
- KIAA1614
- Gene Name
- KIAA1614
There are no transcripts associated with this gene.
- Gene
- STX6
- Gene Name
- syntaxin 6
View Full Product Details