Product Details

SNP ID
rs1980577
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:44753471 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGAGAGTGGAGATGGCTCCTTCCC[A/G]TGGTATCCTGCATGTGATCTTGGCC
Phenotype
MIM: 607368 MIM: 611601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK15 PubMed Links

Gene Details

Gene
KCNK15
Gene Name
potassium two pore domain channel subfamily K member 15
There are no transcripts associated with this gene.

Gene
KCNK15-AS1
Gene Name
KCNK15 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
RIMS4
Gene Name
regulating synaptic membrane exocytosis 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205317.1 3540 UTR 3 NP_001192246.1
NM_182970.3 3540 UTR 3 NP_892015.1

View Full Product Details