Product Details

SNP ID
rs15230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50935001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAATATAAAATAGGTGGTCTT[A/C]ATAAAAAGATGCATGAAATTTACCT
Phenotype
MIM: 611386 MIM: 603503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADNP PubMed Links

Gene Details

Gene
ADNP
Gene Name
activity dependent neuroprotector homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282531.1 955 Intron NP_001269460.1
NM_001282532.1 955 Intron NP_001269461.1
NM_015339.3 955 Intron NP_056154.1
NM_181442.2 955 Intron NP_852107.1
XM_011528747.2 955 Intron XP_011527049.1
XM_011528748.2 955 Intron XP_011527050.1
XM_017027757.1 955 Intron XP_016883246.1
XM_017027758.1 955 Intron XP_016883247.1
XM_017027759.1 955 Intron XP_016883248.1
Gene
ADNP-AS1
Gene Name
ADNP antisense RNA 1
There are no transcripts associated with this gene.

Gene
DPM1
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317034.1 955 UTR 3 NP_001303963.1
NM_001317035.1 955 UTR 3 NP_001303964.1
NM_001317036.1 955 UTR 3 NP_001303965.1
NM_003859.2 955 UTR 3 NP_003850.1
XM_011529093.2 955 Intron XP_011527395.1

View Full Product Details