Product Details

SNP ID
rs12610295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19928158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGTATCATTCAAGAAAATAAGC[A/C]AACTTTATCAGTGTTGATAATCAGT
Phenotype
MIM: 603975
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF93 PubMed Links

Gene Details

Gene
ZNF93
Gene Name
zinc finger protein 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031218.3 Intron NP_112495.2

View Full Product Details