Product Details
- SNP ID
-
rs1977525
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:29541272 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGCCACCGTGTCCGGCCTACAAG[C/T]GACTTTTAATTACAGTTTAGAATTG
- Phenotype
-
MIM: 138245
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRIK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112599701,rs140219021] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRIK1
- Gene Name
- glutamate ionotropic receptor kainate type subunit 1
View Full Product Details