Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001002837.2 | Intron | NP_001002837.1 | |||
NM_001284285.1 | Intron | NP_001271214.1 | |||
NM_001284286.1 | Intron | NP_001271215.1 | |||
NM_001284287.1 | Intron | NP_001271216.1 | |||
NM_001284288.1 | Intron | NP_001271217.1 | |||
NM_001284289.1 | Intron | NP_001271218.1 | |||
XM_011530143.1 | Intron | XP_011528445.1 | |||
XM_017028771.1 | Intron | XP_016884260.1 | |||
XM_017028772.1 | Intron | XP_016884261.1 | |||
XM_017028773.1 | Intron | XP_016884262.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_080430.2 | Intron | NP_536355.1 |