Product Details

SNP ID
rs13105517
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153701798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAGAGGGAATGCGAAGCA[A/G]TGGTTTTGGTGCGAACACAAAAATT
Phenotype
MIM: 603028
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RNF175 PubMed Links

Gene Details

Gene
RNF175
Gene Name
ring finger protein 175
There are no transcripts associated with this gene.

Gene
TLR2
Gene Name
toll like receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318787.1 299 Intron NP_001305716.1
NM_001318789.1 299 Intron NP_001305718.1
NM_001318790.1 299 Intron NP_001305719.1
NM_001318791.1 299 Intron NP_001305720.1
NM_001318793.1 299 Intron NP_001305722.1
NM_001318795.1 299 Intron NP_001305724.1
NM_001318796.1 299 Intron NP_001305725.1
NM_003264.4 299 Intron NP_003255.2
XM_011532215.2 299 Intron XP_011530517.1
XM_011532216.2 299 Intron XP_011530518.1
XM_017008573.1 299 Intron XP_016864062.1
XM_017008574.1 299 Intron XP_016864063.1
XM_017008575.1 299 Intron XP_016864064.1
XM_017008576.1 299 UTR 5 XP_016864065.1

View Full Product Details