Product Details

SNP ID
rs34864313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64892119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTGCATGCCTCCTTTCCTTCCA[A/G]TGAAAAGAAATCACTGGATATTGCA
Phenotype
MIM: 604756
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC441239 PubMed Links

Gene Details

Gene
LOC441239
Gene Name
uncharacterized LOC441239
There are no transcripts associated with this gene.

Gene
ZNF273
Gene Name
zinc finger protein 273
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021148.2 3820 Intron NP_066971.2
XM_011515746.2 3820 Intron XP_011514048.1
XM_017011688.1 3820 Intron XP_016867177.1
XM_017011689.1 3820 UTR 5 XP_016867178.1
XM_017011690.1 3820 UTR 5 XP_016867179.1
XM_017011691.1 3820 Intron XP_016867180.1
XM_017011692.1 3820 UTR 5 XP_016867181.1
XM_017011693.1 3820 Intron XP_016867182.1
XM_017011694.1 3820 Intron XP_016867183.1
XM_017011695.1 3820 Intron XP_016867184.1
XM_017011696.1 3820 Intron XP_016867185.1
XM_017011697.1 3820 UTR 5 XP_016867186.1
XM_017011698.1 3820 Intron XP_016867187.1

View Full Product Details