Product Details

SNP ID
rs10749865
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:46843593 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTATATATGACAACTTTTGTAGG[A/G]AAACAAGATCCATTTTTTTTTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP4Z2P PubMed Links

Gene Details

Gene
CYP4Z2P
Gene Name
cytochrome P450 family 4 subfamily Z member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details