Product Details

SNP ID
rs3860154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121886653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAGTCACGCAGTTTGAGGTAAC[A/T]TTTTAGGCAGCCCTAGGAAACTGAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TTLL11 PubMed Links

Gene Details

Gene
TTLL11
Gene Name
tubulin tyrosine ligase like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139442.1 Intron NP_001132914.1
NM_194252.2 Intron NP_919228.2
XM_005251728.2 Intron XP_005251785.1
XM_005251729.4 Intron XP_005251786.1
XM_006716966.3 Intron XP_006717029.1
XM_017014309.1 Intron XP_016869798.1
Gene
TTLL11-IT1
Gene Name
TTLL11 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details