Product Details
- SNP ID
-
rs11885
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:753919 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCCCGCAGAGTGCTCCACCGGA[G/T]CCCCACCGGGAAGAGACCGTGACCG
- Phenotype
-
MIM: 605232
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WNK1
PubMed Links
Gene Details
- Gene
- WNK1
- Gene Name
- WNK lysine deficient protein kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184985.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001171914.1 |
NM_014823.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
NP_055638.2 |
NM_018979.3 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
NP_061852.3 |
NM_213655.4 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
NP_998820.3 |
XM_006719003.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_006719066.1 |
XM_011520997.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519299.1 |
XM_011520998.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519300.1 |
XM_011520999.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519301.1 |
XM_011521000.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519302.1 |
XM_011521001.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519303.1 |
XM_011521002.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519304.1 |
XM_011521003.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519305.1 |
XM_011521004.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519306.1 |
XM_011521005.2 |
1322 |
Intron |
|
|
XP_011519307.1 |
XM_011521006.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519308.1 |
XM_011521007.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519309.1 |
XM_011521008.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519310.1 |
XM_011521009.2 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_011519311.1 |
XM_017019834.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875323.1 |
XM_017019835.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875324.1 |
XM_017019836.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875325.1 |
XM_017019837.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875326.1 |
XM_017019838.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875327.1 |
XM_017019839.1 |
1322 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016875328.1 |
View Full Product Details