Product Details

SNP ID
rs1042926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33065045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTGAAGTATTTATTGGTCTTAG[A/G]GTATTCTCGGGAAGGTGACAGTGAG
Phenotype
MIM: 142880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-DPA1 PubMed Links

Gene Details

Gene
HLA-DPA1
Gene Name
major histocompatibility complex, class II, DP alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242524.1 1220 UTR 3 NP_001229453.1
NM_001242525.1 1220 UTR 3 NP_001229454.1
NM_033554.3 1220 UTR 3 NP_291032.2

View Full Product Details